Learn More
Hamilton™ Microlab 300 Pipetting System Accessories
Click to view available options
Type:
Controller
Controller Cable
Hand Probe Assembly
O-ring Kit (for Hand Probe Assembly)
Thumb Screw (for Drive Unit only)
Tubing Kit (for Hand Probe)
User Manual
Valve Assembly
Unit Size:
Each
Specifications
Specifications
| Type | Tubing Kit (for Hand Probe) |
| For Use With (Equipment) | Microlab™ 300 pipetteting system |
| Includes | 5 Ferrules, 5 Stainless Steel rings, 5 fittings, 5 strain reliefs, 12G tubing and a probe tip |