Learn More
Hamilton™ Luer Lock Adapters for Hamilton™Valves
Description
Fittings have a 1/4-28 thread connection on one side and either a male luer, female luer, or male luer lock on the other side.
Specifications
Specifications
| Type | Adapter with Female Luer Lock |
| Thread Size | 1/4-28 Thread .059 in. (1.5mm) |