Learn More
Hamilton™ Luer Lock Adapters for Hamilton™Valves
Product Code. 10271543
Change view
Click to view available options
Type:
Adapter with Female Luer Lock
Adapter with Male Luer Lock
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10271543 Adapter with Male Luer Lock Each
10313033 Adapter with Female Luer Lock Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10271543 Supplier Hamilton™ Supplier No. 35030
Adapters are designed to work with Hamilton valves
Fittings have a 1/4-28 thread connection on one side and either a male luer, female luer, or male luer lock on the other side.
Specifications
Type Adapter with Male Luer Lock
Thread Size 1/4-28 Thread .059 in. (1.5mm)