Learn More
Hamilton™ Constant Rate-700 HPLC Syringes & Replacement Parts
Product Code. 11548833
Change view
Click to view available options
Needle Gauge:
22 G
22s G
Product Type:
Gastight Syringe
Replacement Needle
Model:
CR-700
CR700-20
CR700-200
CR700-50
Unit Size:
Each
Pack of 2
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge Product Type Model unitSize
11548833 22 G Replacement Needle CR-700 Each
10055610 22 G Gastight Syringe CR700-200 Each
15789249 22 G Replacement Needle CR-700 Pack of 2
10015590 22s G Gastight Syringe CR700-20 Each
10508122 22 G Gastight Syringe CR700-50 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11548833 Supplier Hamilton™ Supplier No. 15936
Only null left
Add to basket
Add to basket
Syringes are designed to delive reasy, reproducible injection of liquid samples into an injector on HPLC instrumentation. The CR-700 standardizes three major variables: sample volume, rate of injection, and depth of needle penetration. Even when operated by different technicians, the CR-700 provides reproducibility of +1% of the total syringe volume in routine procedures.
Volumes are selected with micrometer precisions, the plunger is spring-driven and injects the samples at a constant rate. Three models of the CR-700 syringes are available with volumes of 20, 50 or 200μL.
Needle Point Styles
  • Point Style 3 - Blunt needle point for use with HPLC injection valves and for sample pipetting. Available gauge: 22
Specifications
Needle Gauge 22 G
Length (English) Needle 2 in.
Product Type Replacement Needle
Model CR-700
Quantity 1 Ea.
Needle Point Style Blunt
For Use With (Equipment) HPLC Injection Valves