Learn More
Hamilton™ Chemically Inert Manual Valves
Product Code. 10337272
Change view
Click to view available options
Type:
1-1 Manual Valve
3-2 Manual Valve
3-3 Manual Valve
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10337272 3-3 Manual Valve Each
11562425 1-1 Manual Valve Each
10217362 3-2 Manual Valve Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10337272 Supplier Hamilton™ Supplier No. 86727
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
Valves have a fluid path containing PTFE and CTFE
The ports on the valve are 1/4-28 threads and come in different flow paths.
Specifications
For Use With (Application) For low pressure applications, from a basic on/off flow to a four-way distribution system
Material CTFE, PTFE
Type 3-3 Manual Valve