Learn More
Hamilton™ Chaney Adapter Assemb 700 800 7000/1700 1800 Series Microliter/Gastight™ Syring
Product Code. 10015600
Change view
Click to view available options
For Use With (Equipment):
Hamilton Syringe Models 702-750, 7000 and 1700 Series
Hamilton Syringe Models 75 and 701 Series
Hamilton Syringe Models 800 and 1800 Series
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10015600 Hamilton Syringe Models 702-750, 7000 and 1700 Series Each
10281961 Hamilton Syringe Models 75 and 701 Series Each
13246949 Hamilton Syringe Models 702-750, 7000 and 1700 Series Each
10343213 Hamilton Syringe Models 800 and 1800 Series Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10015600 Supplier Hamilton™ Supplier No. 14725
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed to work with 700, 800, 7000 series Microliter syringes and 1700 and 1800 series Gastight syringes
Adapters are designed to give consistent and reproducible injections and also prevent the plungers from bending.
Specifications
Product Type Reproducibility Adapter
Quantity Ea.
For Use With (Equipment) Hamilton Syringe Models 702-750, 7000 and 1700 Series