missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Halt™ Protease Inhibitor Cocktail, EDTA-Free (100X)

Product Code. 10444414 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
24 x 100 μL
Unit Size:
10mL
1mL
5mL
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10444414 24 x 100 μL Each
11804111 5 mL 5mL
11824111 10 mL 10mL
10320015 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Item(s) in this group are an alternative option to part number P9599
Product Code. 10444414 Supplier Thermo Scientific™ Supplier No. 78425

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Item(s) in this group are an alternative option to part number P9599

Inhibit proteolytic degradation during cell lysis and protein extraction with these ready-to-use concentrated stock solutions of protease inhibitors.

Thermo Scientific Halt Protease Inhibitor Cocktail, EDTA-free (100X) are ready-to-use concentrated stock solutions of protease inhibitors for addition to samples to prevent proteolytic degradation during cell lysis and protein extraction.

Features of Halt Protease Inhibitor Cocktail, EDTA-free (100X):

  • Convenient—the refrigerator-stable, 100X liquid format is more effective and easier to use than individual inhibitors or other formats; just pipette exactly the amount you need
  • No proprietary ingredients—fully disclosed formulation contains optimized concentrations of six broad-spectrum protease inhibitors AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A stabilized in high-quality dimethylsulfoxide (DMSO)
  • Multiple package sizes—choose among several package sizes, including single-use microtubes that keep protease inhibitors fresh and reduce the risk of reagent contamination
  • EDTA-free formulations—ensures compatibility with isoelectric focusing
  • Lysis buffer compatible—use with Thermo Scientific Pierce Cell Lysis Buffers or nearly any other commercial or homemade detergent-based lysis reagent. Add 10 μL of concentrated cocktail per 1 mL of lysis buffer to ensure complete protection of the resulting protein extract.

Applications:

  • Protection of intact, active cellular proteins from degradation by endogenous proteases
  • Inhibition of protease activity from lysates produced by a variety of methods including Thermo Scientific Pierce Cell Lysis Reagents, other commercially formulated cell or tissue lysis products, sonication, French press, blender homogenization or repetitive freeze/thaw cycling
  • Screening of extracts for proteolytic activity
  • The study of proteolysis in the regulation of cellular processes

Proteases make up a large category of enzymes, including endopeptidases and exopeptidases, which catalyze the hydrolytic breakdown of proteins into peptides or amino acids in a wide variety of cell and tissue types. Proteases are regulated and compartmentalized in living cells, but cell lysis releases and mixes them with other extracted proteins. Their activity can thus lead to the loss of a large number of valuable proteins, adversely affecting downstream applications. For this reason, it is common practice to spike cell lysis buffers or cell extracts with a cocktails of compounds known to inactivate or inhibit proteases. Halt Protease Inhibitor Cocktails effectively inhibit serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. Regular and EDTA-free formulations provide for inhibition of metalloproteases and compatibility with 2D electrophoresis, respectively.

Related Products

  • Halt™ Protease Inhibitor Single-Use Cocktail (100X) (Cat. No. 78430)
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protease Inhibitor Cocktail
Content And Storage Upon receipt store at 4°C
Form Liquid
Product Line Halt
Buffer Lysis Buffer
Reagent Type Protease Inhibitor
Inhibitors Protease Inhibitor
Quantity 24 x 100 μL
compliance-icons
Product Identifier
  • Halt Protease Inhibitor Cocktail
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Benzenesulfonyl fluoride, 4-(2-aminoethyl)-hydrochloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.