missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haloxyfop-R (free acid), Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Haloxyfop-R (free acid) |
| CAS | 95977-29-0 |
| Analyte or Component Names | Haloxyfop-R (free acid) |
| Shipping Condition | +20°C |
| Molecular Formula | C15 H11 Cl F3 N O4 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Propanoic acid, 2-[4-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy]-, (R)-, (2R)-2-[4-[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy]propanoic acid, (+)-Haloxyfop, (R)-(+)-Haloxyfop, (R)-Haloxyfop, Haloxyfop-P |
| InChI Formula | InChI=1S/C15H11ClF3NO4/c1-8(14(21)22)23-10-2-4-11(5-3-10)24-13-12(16)6-9(7-20-13)15(17,18)19/h2-8H,1H3,(H,21,22)/t8-/m1/s1 |
| SMILES | C[C@@H](Oc1ccc(Oc2ncc(cc2Cl)C(F)(F)F)cc1)C(=O)O |
| IUPAC Name | (2R)-2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxyphenoxy]propanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?