missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haloxyfop-methyl-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Haloxyfop-methyl-d4 |
| Molecular Formula | C16H9D4ClF3NO4 |
| Quantity | 50 mg |
| Synonym | 2-[4-[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy]-propanoic Acid Methyl Ester, Dowco 453, Dowco 453ME, Haloxyfop methyl ester, Haloxyfop-Me, Verdict, Verdict R |
| InChI Formula | InChI=1S/C16H13ClF3NO4/c1-9(15(22)23-2)24-11-3-5-12(6-4-11)25-14-13(17)7-10(8-21-14)16(18,19)20/h3-9H,1-2H3/i3D,4D,5D,6D |
| SMILES | ClC(C=C(C(F)(F)F)C=N1)=C1OC2=C([2H])C([2H])=C(OC(C)C(OC)=O)C([2H])=C2[2H] |
| Molecular Weight (g/mol) | 379.75 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?