missing translation for 'onlineSavingsMsg'
Learn More

Haloxyfop (free acid) D4 (phenoxy-D4) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30318262
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30318262 1 mL 1mL
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30318262

Brand: Dr. Ehrenstorfer DREA14060010AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Haloxyfop (free acid) phenoxy-d4
CAS 127893-34-9
Solvent or Matrix Acetonitrile
Molecular Formula C15 2H4 H7 Cl F3 N O4
Quantity 1 mL
Synonym Haloxyfop (Free Acid) D4 (Phenoxy-D4), Propanoic acid, 2-[4-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy-2,3,5,6-d4]- (9CI)
InChI Formula InChI=1S/C15H11ClF3NO4/c1-8(14(21)22)23-10-2-4-11(5-3-10)24-13-12(16)6-9(7-20-13)15(17,18)19/h2-8H,1H3,(H,21,22)/i2D,3D,4D,5D
SMILES [2H]c1c([2H])c(Oc2ncc(cc2Cl)C(F)(F)F)c([2H])c([2H])c1OC(C)C(=O)O
IUPAC Name 2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy-2,3,5,6-tetradeuteriophenoxy]propanoic acid
Molecular Weight (g/mol) 365.72
Formula Weight 365.058 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.