missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Haloxyfop-d4, TRC
Specifications
Specifications
| Chemical Name or Material | (±)-Haloxyfop (free acid) phenoxy-d4 |
| CAS | 127893-34-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 2H4 H7 Cl F3 N O4 |
| Quantity | 2.5 mg |
| Synonym | Haloxyfop (Free Acid) D4 (Phenoxy-D4), Propanoic acid, 2-[4-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy-2,3,5,6-d4]- (9CI), Haloxyfop D4 (phenoxy D4) |
| InChI Formula | InChI=1S/C15H11ClF3NO4/c1-8(14(21)22)23-10-2-4-11(5-3-10)24-13-12(16)6-9(7-20-13)15(17,18)19/h2-8H,1H3,(H,21,22)/i2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c(Oc2ncc(cc2Cl)C(F)(F)F)c([2H])c([2H])c1OC(C)C(=O)O |
| IUPAC Name | 2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy-2,3,5,6-tetradeuteriophenoxy]propanoic acid |
| Molecular Weight (g/mol) | 365.72 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?