missing translation for 'onlineSavingsMsg'
Learn More

(±)-Haloxyfop-d4, TRC

Product Code. 30391308
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391308 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30391308 Supplier Toronto Research Chemicals Supplier No. TRCH10404225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Haloxyfop (free acid) phenoxy-d4
CAS 127893-34-9
Purity Grade Notes HPLC
Molecular Formula C15 2H4 H7 Cl F3 N O4
Quantity 25 mg
Synonym Haloxyfop (Free Acid) D4 (Phenoxy-D4), Propanoic acid, 2-[4-[[3-chloro-5-(trifluoromethyl)-2-pyridinyl]oxy]phenoxy-2,3,5,6-d4]- (9CI), Haloxyfop D4 (phenoxy D4)
InChI Formula InChI=1S/C15H11ClF3NO4/c1-8(14(21)22)23-10-2-4-11(5-3-10)24-13-12(16)6-9(7-20-13)15(17,18)19/h2-8H,1H3,(H,21,22)/i2D,3D,4D,5D
SMILES [2H]c1c([2H])c(Oc2ncc(cc2Cl)C(F)(F)F)c([2H])c([2H])c1OC(C)C(=O)O
IUPAC Name 2-[4-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy-2,3,5,6-tetradeuteriophenoxy]propanoic acid
Molecular Weight (g/mol) 365.72
Formula Weight 365.058
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.