missing translation for 'onlineSavingsMsg'
Learn More

Haloperidol, MedChemExpress

Product Code. 16481726
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481726 500 mg 500mg
16436486 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481726 Supplier Medchem Express Supplier No. HY14538
 Restricted Item
Estimated Shipment: 23-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Haloperidol is a potent dopamine D2 receptor antagonist, widely used as an antipsychotic.

Haloperidol is a potent dopamine D2 receptor antagonist, widely used as an antipsychotic.

Specifications

Chemical Name or Material Haloperidol
CAS 52-86-8
Color White
Purity Grade Notes Research
Molecular Formula C21H23ClFNO2
Quantity 500 mg
Solubility Information DMSO : 16.67 mg/mL (44.35 mM; Need ultrasonic)
SMILES ClC(C=C1)=CC=C1C2(O)CCN(CCCC(C3=CC=C(F)C=C3)=O)CC2
Molecular Weight (g/mol) 375.86
Formula Weight 375.86
Percent Purity 99.36%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.