missing translation for 'onlineSavingsMsg'
Learn More

Halofuginone hydrobromide, Dr. Ehrenstorfer

Product Code. 30320704
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320704 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30320704 Supplier Dr. Ehrenstorfer Supplier No. DREC14059280
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Halofuginone Hydrobromide
CAS 64924-67-0
Analyte or Component Names Halofuginone Hydrobromide
Shipping Condition +20°C
Molecular Formula C16 H17 Br Cl N3 O3 . Br H
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C16H17BrClN3O3.BrH/c17-11-6-13-10(5-12(11)18)16(24)21(8-20-13)7-9(22)4-14-15(23)2-1-3-19-14;/h5-6,8,14-15,19,23H,1-4,7H2;1H/t14-,15+;/m0./s1
SMILES Br.O[C@@H]1CCCN[C@H]1CC(=O)CN2C=Nc3cc(Br)c(Cl)cc3C2=O
IUPAC Name 7-bromo-6-chloro-3-[3-[(2S,3R)-3-hydroxypiperidin-2-yl]-2-oxopropyl]quinazolin-4-one;hydrobromide
Molecular Weight (g/mol) 495.59
Quantity 10 mg
Formula Weight 492.9403 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.