missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Halofantrine hydrochloride, MedChemExpress
Description
Halofantrine hydrochloride (SKF-102886) is a blocker of delayed rectifier potassium current via the inhibition of human-ether-a-go-go-related gene (HERG) channel and a potent antimalarial compound.
Specifications
Specifications
| Chemical Name or Material | Halofantrine hydrochloride |
| Physical Form | Solid |
| CAS | 36167-63-2 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C26H31Cl3F3NO |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | SKF-102886WR-171669 hydrochloride |
| Solubility Information | DMSO : 30 mg/mL (55.88 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?