missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Halobetasol Propionate, TRC
Specifications
Specifications
| Chemical Name or Material | Halobetasol Propionate |
| CAS | 66852-54-8 |
| Molecular Formula | C25 H31 Cl F2 O5 |
| Quantity | 100 mg |
| Synonym | (6α,11β,16β)-21-Chloro-6,9-difluoro-11-hydroxy-16-methyl-17-(1-oxopropoxy)-pregna-1,4-diene-3,20-dione, BMY 30056, CGP 14458, Halobetasol propionate, Miracorten, Ulobetasol propionate, Ultravate, (6α,11β,16β)-21-Chloro-6,9-difluoro-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione 17-propionate |
| InChI Formula | InChI=1S/C25H31ClF2O5/c1-5-21(32)33-25(20(31)12-26)13(2)8-15-16-10-18(27)17-9-14(29)6-7-22(17,3)24(16,28)19(30)11-23(15,25)4/h6-7,9,13,15-16,18-19,30H,5,8,10-12H2,1-4H3/t13-,15-,16-,18-,19-,22-,23-,24-,25-/m0/s1 |
| SMILES | CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CCl |
| IUPAC Name | [(6S,8S,9R,10S,11S,13S,14S,16S,17R)-17-(2-chloroacetyl)-6,9-difluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate |
| Molecular Weight (g/mol) | 484.96 |
| Formula Weight | 484.1828 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?