missing translation for 'onlineSavingsMsg'
Learn More

Halobetasol (propionate), MedChemExpress

Product Code. 16494497
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494497 50 mg 50mg
16458476 100 mg 100mg
16438638 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494497 Supplier Medchem Express Supplier No. HYB0878

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Halobetasol propionate is a synthetic corticosteroid for topical dermatological use; exhibits anti-inflammatory, antipruritic, and vasoconstrictive properties.

Halobetasol propionate is a synthetic corticosteroid for topical dermatological use

Specifications

Chemical Name or Material Halobetasol (propionate)
CAS 66852-54-8
Color White
Purity Grade Notes Research
Molecular Formula C25H31ClF2O5
Quantity 50 mg
Synonym BMY-30056 CGP-14458 Ulobetasol propionate
Solubility Information DMSO : ≥ 100 mg/mL (206.20 mM)
SMILES C[C@@]12[C@](C(CCl)=O)(OC(CC)=O)[C@@H](C)C[C@@]1([H])[C@]3([H])C[C@H](F)C4=CC(C=C[C@]4(C)[C@@]3(F)[C@@H](O)C2)=O
Molecular Weight (g/mol) 484.96
Formula Weight 484.96
Percent Purity 99.91%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.