missing translation for 'onlineSavingsMsg'
Learn More

Miele™ Half Mesh Basket

Brand:  Miele™ 3830300

Code : NEW

Additional Details : Weight : 2.50000kg

Product Code. 15253568

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

Mesh tray, 1mm wire mesh, 5mm mesh size, 5mm all-round frame, 2 hinged handles
upper basket O 191/1
45/55 mm
10 kg
Half Mesh Basket
255 mm
480 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers