Learn More
Eppendorf™ Half Height Freezer Rack for Chest Freezers
Store your freezer storage boxes within organized freezer racks. These metal racks optimize your freezer space and make it easy to find the items you desire.
Brand: Eppendorf™ K0641-306A
165.00 GBP valid until 2024-12-31
Use promo code "23275" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.20000kg
Description
- Half Height Freezer Rack for Chest Freezers
- Available in a variety of sizes
Specifications
Half Height Freezer Rack | |
2 in. Boxes (7) | |
Chest Freezer |