missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Demi-Fraser Broth (without Ferric Ammonium Citrate) (Dehydrated)

Product Code. 12864452
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Unit Size:
10kg
3kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12864452 10 kg 10kg
11903882 500 g 500g
12874452 2.5 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12864452 Supplier Thermo Scientific™ Supplier No. CM1053G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate Listeria spp. from food and environmental samples with this medium.

Isolate Listeria spp. from food and environmental samples with Thermo Scientific™ Demi-Fraser Broth (without Ferric Ammonium Citrate) (Dehydrated), when used as the secondary enrichment medium in methodology for Listeria isolation. This medium is a modification of Fraser Broth and contains half of the concentration of nalidixic acid and acriflavin hydrochloride to aid in the recovery of stressed cells. The removal of ferric ammonium citrate from the formula aids in subculture from broth to imaging instrumentation or Polymerase Chain Reaction-based assay machines.

Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both humans and animals. Use Demi-Fraser Broth (without Ferric Ammonium Citrate) for isolation of Listeria species from food and environmental samples when used as the secondary enrichment medium in methodology for Listeria isolation.

  • Helps in the recovery of stressed cells
  • Allows subculture from the broth to imaging instrumentation or PCR based assay machines

Not all products are available for sale in all territories. Please inquire.

Remel™ and Thermo Scientific™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Additive Without Ferric Ammonium Citrate
Media Type Demi-Fraser Broth
Product Type Nutrient Broth
For Use With (Application) Pre-supplemented primary selective enrichment broth for &italic_begin;Listeria&italic_end; spp.
Quantity 10 kg
Yield 174.4 L medium

For Laboratory Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.