missing translation for 'onlineSavingsMsg'
Learn More

Halcinonide, TRC

Product Code. 30396266
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396266 50 mg
30396267 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396266 Supplier Toronto Research Chemicals Supplier No. TRCH10200050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3093-35-4
Molecular Formula C24 H32 Cl F O5
Molecular Weight (g/mol) 454.96
Synonym Pregn-4-ene-3,20-dione, 21-chloro-9-fluoro-11β,16α,17-trihydroxy-, cyclic 16,17-acetal with acetone (7CI,8CI), (11β,16α)-21-Chloro-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregn-4-ene-3,20-dione, 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregn-4-ene-3,20-dione deriv., Adcortin, Alcinonide, Halciderm, Halcimat, Halcinonide, Halog, SQ 18566
IUPAC Name (11β,16α)-21-Chloro-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregn-4-ene-3,20-dione
SMILES CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)CC[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CCl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C24H32ClFO5/c1-20(2)30-19-10-16-15-6-5-13-9-14(27)7-8-21(13,3)23(15,26)17(28)11-22(16,4)24(19,31-20)18(29)12-25/h9,15-17,19,28H,5-8,10-12H2,1-4H3/t15-,16-,17-,19+,21-,22-,23-,24+/m0/s1
Formula Weight 454.1922
Chemical Name or Material Halcinonide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.