missing translation for 'onlineSavingsMsg'
Learn More

Halcinonide, MedChemExpress

Product Code. 16484497
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484497 50 mg 50mg
16461446 200 mg 200mg
16448476 100 mg 100mg
16428638 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484497 Supplier Medchem Express Supplier No. HYB0877

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Halcinonide (SQ-18566) is a high potency corticosteroid used topically in the treatment of certain skin conditions.

Halcinonide (SQ-18566) is a high potency corticosteroid used topically in the treatment of certain skin conditions.

Specifications

Chemical Name or Material Halcinonide
CAS 3093-35-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H32ClFO5
Quantity 50 mg
Synonym SQ-18566
Solubility Information DMSO : ≥ 48 mg/mL (105.50 mM)
SMILES C[C@@]12[C@@]3(C(CCl)=O)[C@@](OC(C)(O3)C)([H])C[C@@]1([H])[C@]4([H])CCC5=CC(CC[C@]5(C)[C@@]4(F)[C@@H](O)C2)=O
Molecular Weight (g/mol) 454.96
Formula Weight 454.96
Percent Purity 99.87%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.