missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAL 293T Cell Overexpression Lysate (Denatured), Abnova
Description
Western Blotting
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | HAL 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC096097.3 |
| Molecular Weight (g/mol) | 72.7 |
| pH Range | 6.8 |
| Plasmid | pCMV-HAL full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-HAL antibody (H00003034-B01) by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
Safety and Handling
Product Identifier
- 100UL HAL 293T Cell Transient Overexpression Lysat
- Warning
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- MIXTURE LIST-Contain: Mercaptoethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?