missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hairy (Drosophila) Antibody (1/24), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76659JF549
This item is not returnable.
View return policy
Description
Hairy (Drosophila) Monoclonal antibody specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Hairy (Drosophila) | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 2023-01-24 00:00:00 | |
| 50mM Sodium Borate | |
| Hairy gene product | |
| 0.1 mL | |
| Cellular Signaling, Developmental Biology, Epigenetics, Stem Cells | |
| Drosophila | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction