missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Hairy (Drosophila) Monoclonal specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Hairy (Drosophila) |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 43489 |
| Conjugate | HRP |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | PBS with no preservative |
| Host Species | Mouse |
| Immunogen | Hairy gene product |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?