missing translation for 'onlineSavingsMsg'
Learn More

Hairy (Drosophila) Antibody (1/24), DyLight 405, Novus Biologicals™

Product Code. 18333971 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18333971 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18333971 Supplier Novus Biologicals Supplier No. NBP276659V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hairy (Drosophila) Monoclonal specifically detects Hairy (Drosophila) in Drosophila samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hairy (Drosophila)
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 1/24
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen Hairy gene product
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Cellular Signaling, Developmental Biology, Epigenetics, Stem Cells
Primary or Secondary Primary
Test Specificity This antibody will stain Drosophila embryos.
Target Species Drosophila
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.