missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hairless Antibody (OTI1D9), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72023PE
This item is not returnable.
View return policy
Description
Hairless Monoclonal antibody specifically detects Hairless in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Hairless | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 55806 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1D9 | |
| Western Blot, Immunohistochemistry | |
| ALUNC, AUHSA277165, FLJ98880, hairless (mouse) homolog, hairless homolog (mouse), MUHH, MUHH1, protein hairless | |
| Human recombinant protein fragment corresponding to amino acids 946-1189 of human HR(NP_005135) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction