missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAI-2/SPINT2 Rat anti-Mouse, Clone: RM0089-7F52, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-21728
This item is not returnable.
View return policy
Description
HAI-2/SPINT2 Monoclonal specifically detects HAI-2/SPINT2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HAI-2/SPINT2 | |
| Monoclonal | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reconstitute with 0.2 mL sterilized PBS to a final concentration of 0.5 mg/mL. | |
| Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles. | |
| IgG2 |
| Western Blot | |
| RM0089-7F52 | |
| Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2 | |
| Mouse recombinant of the common extracellular region of HAI-2 isoforms | |
| 0.1 mg | |
| Cancer | |
| 10653 | |
| Mouse | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction