missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAI-2/SPINT2 Antibody (RM0089-7F52), Alexa Fluor™ 350, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-21728AF350
This item is not returnable.
View return policy
Description
HAI-2/SPINT2 Monoclonal specifically detects HAI-2/SPINT2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HAI-2/SPINT2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SPINT2 | |
| Mouse recombinant of the common extracellular region of HAI-2 isoforms | |
| 0.1 mL | |
| 10653 | |
| Mouse | |
| IgG2 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0089-7F52 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| This antibody detects mouse HIA-2, but not HAI-1 in Western blotting. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur