missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAI-1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15036-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms.Specifications
| HAI-1 | |
| Polyclonal | |
| Unconjugated | |
| SPINT1 | |
| HAI, HAI 1, HAI1, MANSC2, SPINT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20732, 6692 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43278, Q9R097 | |
| SPINT1 | |
| HAI-1 Fusion Protein Ag6665 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction