missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HAI-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581039
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human HAI-1/SPINT1.
The protein encoded by this gene is a member of the Kunitz family of serine protease inhibitors. The protein is a potent inhibitor specific for HGF activator and is thought to be involved in the regulation of the proteolytic activation of HGF in injured tissues. Alternative splicing results in multiple variants encoding different isoforms.
Specifications
| HAI-1 | |
| Polyclonal | |
| PBS with no preservative | |
| O43278 | |
| SPINT1 | |
| Recombinant Human HAI-1/SPINT1 protein (Met1-Val433). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| SPINT1 | |
| HAI; HAI1; HAI-1; hepatocyte growth factor activator inhibitor 1; hepatocyte growth factor activator inhibitor type 1; hepatocyte growth factor activator inhibitor-1; kunitz-type protease inhibitor 1; MANSC2; serine peptidase inhibitor, Kunitz type 1; serine protease inhibitor, Kunitz type 1; Spint1; UNQ223/PRO256 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6692 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction