missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAI-1/HGFA Inhibitor 1 Antibody (RM0088-3G21), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-12356G
This item is not returnable.
View return policy
Description
HAI-1/HGFA Inhibitor 1 Monoclonal specifically detects HAI-1/HGFA Inhibitor 1 in Mouse samples. It is validated for Western Blot.
Specifications
| HAI-1/HGFA Inhibitor 1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SPINT1 | |
| Mouse recombinant HAI-1/HGFA Inhibitor 1 (extracellular domain) | |
| 0.1 mL | |
| 6692 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| RM0088-3G21 | |
| Western Blot | |
| HAI, HAI1, HAI-1, hepatocyte growth factor activator inhibitor 1, Hepatocyte growth factor activator inhibitor type 1, kunitz-type protease inhibitor 1, MANSC2, serine peptidase inhibitor, Kunitz type 1, serine protease inhibitor, Kunitz type 1 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction