Learn More
Hafnium trifluoromethanesulfonate, 98%
Description
Hafnium trifluoromethanesulfonate is a recyclable catalyst for the mononitration of o-nitrotoluene. It serves as a catalyst for homogeneous methoxycarbonylation and hydrocarboxylation reactions of phenylacetylene. It is also used as a reactant for direct Friedel-Crafts reactions of chromene hemiacetals, aminomethylation reactions under Lewis acidic conditions and prins-type cyclization reactions. It is also involved in the direct polycondensation of lactic acid. Further, it is used in cationic benzylation reactions, chemoselective thioacetalization and transthioacetalization of carbonyl compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Hafnium trifluoromethanesulfonate is a recyclable catalyst for the mononitration of o-nitrotoluene. It serves as a catalyst for homogeneous methoxycarbonylation and hydrocarboxylation reactions of phenylacetylene. It is also used as a reactant for direct Friedel-Crafts reactions of chromene hemiacetals, aminomethylation reactions under Lewis acidic conditions and prins-type cyclization reactions. It is also involved in the direct polycondensation of lactic acid. Further, it is used in cationic benzylation reactions, chemoselective thioacetalization and transthioacetalization of carbonyl compounds.
Solubility
Soluble in methyl cyanide, dicholomethane, dichloroethane, nitromethane, dioxane, terahydrofuran and toluene.
Notes
Incompatible with strong oxidizing agents. Moisture sensitive.
Specifications
Specifications
| Melting Point | >350°C |
| Linear Formula | (CF3SO3)4Hf |
| Quantity | 25 g |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in methyl cyanide,dicholomethane,dichloroethane,nitromethane,dioxane,terahydrofuran and toluene. |
| Formula Weight | 774.75 |
| Percent Purity | 98% |
| Chemical Name or Material | Hafnium trifluoromethanesulfonate |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.