missing translation for 'onlineSavingsMsg'
Learn More

Hafnium tert-butoxide, 99.9% (metals basis excluding Zr), Zr< 0.5%

Product Code. 11304887
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304887 2 g 2g
11394877 10 g 10g
11314887 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11304887 Supplier Thermo Scientific Alfa Aesar Supplier No. 041908.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2172-02-3 | C16H40HfO4 | 474.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2172-02-3
MDL Number MFCD00070458

Specifications

Melting Point 2°C
Color Yellow
Density 1.15 g/mL
Boiling Point 90°C (5 mmHg)
Flash Point 28°C (82°F)
Molecular Formula C16H40HfO4
Linear Formula Hf[OC(CH3)3]4
Quantity 2 g
UN Number UN1993
Sensitivity Air and moisture sensitive
Synonym tetrakis tert-butoxy hafnium, hafnium; 2-methylpropan-2-ol
Solubility Information Soluble in hydrocarbons, reacts with alcohols, ketones, and esters
InChI Key YZABPPRMRYHJGL-UHFFFAOYSA-N
SMILES CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.CC(C)(C)O.[Hf]
IUPAC Name hafnium;2-methylpropan-2-ol
Molecular Weight (g/mol) 474.98
PubChem CID 25199745
Formula Weight 470.94
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis excluding Zr), Zr < 0.5%
Chemical Name or Material Hafnium tert-butoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.