missing translation for 'onlineSavingsMsg'
Learn More

Hafnium standard solution, for AAS, 1 mg/ml Hf in 5% HF

Product Code. 10753581 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10753581 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10753581 Supplier Thermo Scientific Acros Supplier No. 196011000
  • ONLINE EXCLUSIVE PRICE
    £28.75
    £26.80 / 100mL
    Save £1.95
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-58-6 | Hf | 178.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hafnium standard solution
Name Note 1 mg/ml Hf in 5% HF
Packaging Plastic bottle
CAS 7440-58-6, 7664-39-3, 7732-18-5
Physical Form Solution
Molecular Formula Hf
Concentration 990 to 1010 μg/mL of Hf(4+) in 5% HF
MDL Number MFCD00011032
Color Yellow
Solubility Soluble in water
InChI Key VBJZVLUMGGDVMO-UHFFFAOYSA-N
SMILES [Hf]
IUPAC Name hafnium
Molecular Weight (g/mol) 178.49
PubChem CID 23986
Quantity 100 mL
ChEBI CHEBI:33343
Formula Weight 178.49
Grade AAS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hafnium standard solution, 1 mg/ml Hf in 5% HF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H310-Fatal in contact with skin.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.