missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) sulfate, 99.9% (metals basis excluding Zr), Zr <1%

Product Code. 11399768
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399768 5 g 5g
11309778 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11399768 Supplier Thermo Scientific Alfa Aesar Supplier No. 044244.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 15823-43-5 | HfO4S | 274.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15823-43-5
Molecular Formula HfO4S
Molecular Weight (g/mol) 274.55
MDL Number MFCD00168081
InChI Key KOELDGKLEFTHGN-UHFFFAOYSA-L
Synonym hafnium sulfate, acmc-20aksa, sulfuric acid, hafnium 4+ salt 2:1, hafnium 4+ disulfate, hafnium iv sulfate tetrahydrate, sulfuric acid,hafnium 4+ salt 2:1, hafnium iv sulfate trace metals basis purity excludes zirconium
PubChem CID 13094115
IUPAC Name hafnium(4+);disulfate
SMILES [Hf].[O-]S([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 5 g
Formula Weight 370.62 (Anhydrous)
Percent Purity 99.9%
Assay (metals basis excluding Zr), Zr <1%
Chemical Name or Material Hafnium (IV) sulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.