missing translation for 'onlineSavingsMsg'
Learn More

Hafnium(IV) 2,4-pentanedionate, 97%

Product Code. 11368978
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368978 5 g 5g
11358978 1 g 1g
11378978 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11368978 Supplier Thermo Scientific Alfa Aesar Supplier No. 042463.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 17475-67-1 | C20H28HfO8 | 574.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17475-67-1
Molecular Formula C20H28HfO8
Molecular Weight (g/mol) 574.92
MDL Number MFCD00044997
InChI Key GXNMJQUZCICHBX-UHFFFAOYSA-N
Synonym hafnium iv acetylacetonate, hafnium-2,4-pentanedionate, hafnium; z-4-hydroxypent-3-en-2-one, 3z-4-tris 2z-4-oxopent-2-en-2-yl oxy hafnio oxy pent-3-en-2-one
PubChem CID 11330624
IUPAC Name hafnium;(Z)-4-hydroxypent-3-en-2-one
SMILES [Hf+4].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O

Specifications

Melting Point 193°C
Odor Characteristic
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 574.93
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Hafnium (IV) 2,4-pentanedionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.