missing translation for 'onlineSavingsMsg'
Learn More

Hafnium ethoxide, 99.9% (metals basis)

Product Code. 11348748
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348748 25 g 25g
11338748 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11348748 Supplier Thermo Scientific Alfa Aesar Supplier No. 041607.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Preparation of refractory coatings | CAS: 13428-80-3 | C8H24HfO4 | 362.766 g/mol

Hafnium ethoxide is a chemical used in the preparation of refractory coatings. Some of the other applications include Nano-materials & Thin Films, Display & Imaging, Catalysis & Synthesis, Hybrid & Disperse Materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13428-80-3
Molecular Formula C8H24HfO4
Molecular Weight (g/mol) 362.766
MDL Number MFCD00070459
InChI Key ZSVBAHMGGMMOSD-UHFFFAOYSA-N
Synonym hafnium tetraethoxide, hafnium iv ethoxide
PubChem CID 72429791
IUPAC Name ethanol;hafnium
SMILES CCO.CCO.CCO.CCO.[Hf]

Specifications

Melting Point 178°C to 180°C
Boiling Point 180°C to 200°C (13 mmHg)
Quantity 25 g
UN Number UN3180
Sensitivity Moisture sensitive
Solubility Information It is soluble in hydrocarbons,reacts with ketones,esters. Reacts exothermally with water
Formula Weight 358.73
Percent Purity 99.9%
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Hafnium ethoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.