missing translation for 'onlineSavingsMsg'
Learn More

Hafnium dichloride oxide octahydrate, Puratronic™, 99.998% (metals basis excluding Zr), Zr <1%, Thermo Scientific™

Product Code. 11380969
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380969 25 g 25g
11390969 1 g 1g
11300979 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11380969 Supplier Thermo Scientific Alfa Aesar Supplier No. 010705.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14456-34-9
Molecular Formula Cl2H16HfO9
Molecular Weight (g/mol) 409.51
MDL Number MFCD00150417
InChI Key FXQGUVLIMYOGDZ-UHFFFAOYSA-L
Synonym hafnium iv dichloride oxide octahydrate 98+%-hf zr
PubChem CID 131675691
IUPAC Name hafnium;dichloride;nonahydrate
SMILES O.O.O.O.O.O.O.O.Cl[Hf](Cl)=O

Specifications

Color White
Physical Form Powder
Quantity 25 g
Linear Formula HfCl2O·8H2O
UN Number UN3260
Solubility Information Soluble in water.
Formula Weight 409.52 (265.40 Anhydrous)
Percent Purity 99.998%
Grade Puratronic
Odor Pungent
Sensitivity Hygroscopic
Density 2.2 g/mL
Assay (metals basis excluding Zr), Zr <1%
Chemical Name or Material Hafnium dichloride oxide octahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.