missing translation for 'onlineSavingsMsg'
Learn More

Hafnium dichloride oxide octahydrate, 98+% (metals basis excluding Zr), Zr <1.5%

Product Code. 11311888
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11311888

Brand: Thermo Scientific Alfa Aesar 011833.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

For Synthesis of Hafnium (IV) Oxide | CAS: 14456-34-9 | Cl2H16HfO9 | 409.51 g/mol

Hafnium dichloride oxide octahydrate is used in the synthesis of hafnium(IV) oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14456-34-9
Molecular Formula Cl2H16HfO9
Molecular Weight (g/mol) 409.51
MDL Number MFCD00150417
InChI Key FXQGUVLIMYOGDZ-UHFFFAOYSA-L
Synonym hafnium iv dichloride oxide octahydrate 98+%-hf zr
PubChem CID 131675691
IUPAC Name hafnium;dichloride;nonahydrate
SMILES O.O.O.O.O.O.O.O.Cl[Hf](Cl)=O

Specifications

Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range >98% (Metals basis excluding Zr)
Solubility Information Soluble in water.
Formula Weight 409.52 (265.40 Anhydrous)
Odor Pungent
Sensitivity Hygroscopic
Density 2.2
Chemical Name or Material Hafnium dichloride oxide octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.