missing translation for 'onlineSavingsMsg'
Learn More

Hafnium boride, 99.5% (metals basis excluding Zr), Zr <2%, Thermo Scientific Chemicals

Catalog Number 11375887
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
12007-23-7
B2Hf
MFCD00016125
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
12007-23-7
B2Hf
MFCD00016125

Hafnium boride is used in wear resistant coatings. It is also employed as a nuclear reactor control rod. Furthermore, it is used in hypervelocity re-entry vehicles like ICBM heat shields or aerodynamic leading-edges. It is useful in the preparation of high temperature composites in conjunction with silicon carbide (SiC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 3250°C
Physical Form Powder
Quantity 10 g
Linear Formula HfB2
Solubility Information Insoluble in water.
Formula Weight 200.11
Percent Purity 99.5%
Odor Odorless
Density 10.5 g/mL
Assay (metals basis excluding Zr), Zr <2%
Chemical Name or Material Hafnium boride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'