missing translation for 'onlineSavingsMsg'
Learn More

Hafnium boride, 99.5% (metals basis excluding Zr), Zr <2%, Thermo Scientific Chemicals

Product Code. 11365887
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365887 50 g 50g
11375887 10 g 10g
11385887 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11365887 Supplier Thermo Scientific Alfa Aesar Supplier No. 043664.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Hafnium boride is used in wear resistant coatings. It is also employed as a nuclear reactor control rod. Furthermore, it is used in hypervelocity re-entry vehicles like ICBM heat shields or aerodynamic leading-edges. It is useful in the preparation of high temperature composites in conjunction with silicon carbide (SiC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-23-7
Molecular Formula B2Hf
MDL Number MFCD00016125

Specifications

Melting Point 3250°C
Physical Form Powder
Quantity 50 g
Linear Formula HfB2
Solubility Information Insoluble in water.
Formula Weight 200.11
Percent Purity 99.5%
Odor Odorless
Density 10.5 g/mL
Assay (metals basis excluding Zr), Zr <2%
Chemical Name or Material Hafnium boride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.