missing translation for 'onlineSavingsMsg'
Learn More

Haemophilus influenzae LPS Antibody (12D9) - BSA Free, Novus Biologicals™

Product Code. 18335451 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335451 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335451 Supplier Novus Biologicals Supplier No. NBP275002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Haemophilus influenzae LPS Monoclonal specifically detects Haemophilus influenzae LPS in Bacteria samples. It is validated for N/A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Haemophilus influenzae LPS
Classification Monoclonal
Clone 12D9
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation PBS with 0.02% Sodium Azide
Gene Alias H. influenzae Lipopolysaccharide, H. influenzae LPS, Haemophilus influenzae Lipopolysaccharide
Host Species Mouse
Immunogen LPS epitope, TEPC-15
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology, Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.