missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HADHA Monoclonal Antibody (24GB6655)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA557421
This item is not returnable.
View return policy
Description
HADHA Monoclonal Antibody for Western Blot
This gene encodes the alpha subunit of the mitochondrial trifunctional protein, which catalyzes the last three steps of mitochondrial beta-oxidation of long chain fatty acids. The mitochondrial membrane-bound heterocomplex is composed of four alpha and four beta subunits, with the alpha subunit catalyzing the 3-hydroxyacyl-CoA dehydrogenase and enoyl-CoA hydratase activities. Mutations in this gene result in trifunctional protein deficiency or LCHAD deficiency. The genes of the alpha and beta subunits of the mitochondrial trifunctional protein are located adjacent to each other in the human genome in a head-to-head orientation.
Specifications
| HADHA | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P40939, Q64428, Q8BMS1 | |
| HADHA | |
| Recombinant protein of human HADHA (aa 106-325). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| 24GB6655 | |
| Unconjugated | |
| HADHA | |
| 3-ketoacyl-Coenzyme A (CoA) thiolase, alpha subunit; 3-oxoacyl-CoA thiolase; 78 kDa gastrin-binding protein; C77020; ECHA; gastrin-binding protein; GBP; HADH; HADHA; hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), alpha subunit; hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme A thiolase/enoyl-Coenzyme A hydratase (trifunctional protein), alpha subunit; LCEH; LCHAD; Long chain 3-hydroxyacyl-CoA dehydrogenase; long-chain 2-enoyl-CoA hydratase; Long-chain enoyl-CoA hydratase; long-chain-3-hydroxyacyl-CoA dehydrogenase; mitochondrial long-chain 2-enoyl-Coenzyme A (CoA) hydratase, alpha subunit; mitochondrial long-chain L-3-hydroxyacyl-Coenzyme A (CoA) dehydrogenase, alpha subunit; mitochondrial trifunctional enzyme, alpha subunit; mitochondrial trifunctional protein, alpha subunit; Monolysocardiolipin acyltransferase; Mtpa; RGD1560655; TP-alpha; Trifunctional enzyme subunit alpha, mitochondrial | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 170670, 3030, 97212 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction