missing translation for 'onlineSavingsMsg'
Learn More

HADH, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Rabbit Polyclonal Antibody

Brand:  Abnova H00003033-D01.100uL

Product Code. 16178637

  • £316.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: MAFVTRQFMRSVSSSSTASASAKKIIVKHVTVIGGGLMGAGIAQVAAATGHTVVLVDQTEDILAKSKKGIEESLRKVAKKKFAENPKAGDEFVEKTLSTIATSTDAASVVHSTDLVVEAIVENLKVKNELFKRLDKFAAEHTIFASNTSSLQITSIANATTRQDRFAGLHFFNPVPVMKLVEVIKTPMTSQKTFESLVDFSKALGKHPVSCKDTPGFIVNRLLVPYLMEAIRLYERGDASKEDIDTAMKLGAGYPMGPFELLDYVGLDTTKFIVDGWHEMDAENPLHQPSPSLNKLVAENKFGKKTGEGFYKYK
Specifications

Specifications

HADH
Polyclonal
No additive
BC000306.2
HADH
HADH (AAH00306.1, 1 a.a. ∼ 314 a.a) full-length human protein.
RUO
3033
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
hydroxyacyl-Coenzyme A dehydrogenase
HAD/HADH1/HADHSC/HHF4/M/SCHAD/MGC8392/SCHAD
Rabbit
100 μL
Primary
Human, Mouse
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.