Learn More
Invitrogen™ HADH Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528203
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, GL261, C8D30, PC-12. Predicted reactivity: Mouse (92%), Rat (92%), Xenopus laevis (82%), Pig (95%), Chicken (80%), Rhesus Monkey (98%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the 3-hydroxyacyl-CoA dehydrogenase gene family. The encoded protein functions in the mitochondrial matrix to catalyze the oxidation of straight-chain 3-hydroxyacyl-CoAs as part of the beta-oxidation pathway. Its enzymatic activity is highest with medium-chain-length fatty acids. Mutations in this gene cause one form of familial hyperinsulinemic hypoglycemia. The human genome contains a related pseudogene of this gene on chromosome 15.
Specifications
| HADH | |
| Polyclonal | |
| Unconjugated | |
| HADH | |
| AA409008; AU019341; AW742602; Had; HADH; HADH1; HADHSC; HCDH; HHF4; hydroxyacyl-CoA dehydrogenase; hydroxyacyl-Coenzyme A dehydrogenase; hydroxyacyl-coenzyme A dehydrogenase, mitochondrial; hydroxylacyl-Coenzyme A dehydrogenase short chain; hydroxylacyl-Coenzyme A dehydrogenase, short chain; hydroxylacyl-Coenzyme A dehydrogenase-dehydrogenase; L-3-hydroxyacyl-CoA dehydrogenase; L-3-hydroxyacyl-Coenzyme A dehydrogenase, short chain; M/SCHAD; medium and short chain L-3-hydroxyacyl-coenzyme A dehydrogenase; medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase; MGC8392; Mschad; SCHAD; short chain 3-hydroxyacyl-CoA dehydrogenase; short-chain 3-hydroxyacyl-CoA dehydrogenase; testis secretory sperm-binding protein Li 203a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 113965, 15107, 3033 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q16836, Q61425, Q9WVK7 | |
| HADH | |
| Recombinant protein encompassing a sequence within the center region of human HADH. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.