missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HADH Antibody (D10-E7-R), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-32416
This item is not returnable.
View return policy
Description
HADH Monoclonal antibody specifically detects HADH in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| HADH | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:150, Immunohistochemistry-Paraffin 1:200-1:1:1000 | |
| EC 1.1.1, EC 1.1.1.35, HADH1, HADHSChydroxyacyl-Coenzyme A dehydrogenase, HCDH, hydroxyacyl-CoA dehydrogenase, L-3-hydroxyacyl-Coenzyme A dehydrogenase, short chain, Medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase, MGC8392, mitochondrial, MSCHAD, SCHADHHF4 | |
| Synthetic peptide within Human HADH aa 265-314/ 314. (Uniprot: Q16836) | |
| 100 μL | |
| Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
| 3033 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| D10-E7-R | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction