Learn More
HACL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15519-1-AP-150UL
 View more versions of this product
Product Code. 16864724
  • £349.00 / 150µL
Description
HACL1 gene ontology annotations related to this gene include lyase activity; magnesium ion binding; thiamine pyrophosphate binding.
Specifications
Specifications
HACL1
Polyclonal
Unconjugated
HACL1
2 HPCL, 2 hydroxyacyl CoA lyase 1, 2 hydroxyphytanoyl CoA lyase, HACL1, HPCL, HPCL2, PHYH2, Phytanoyl CoA 2 hydroxylase 2
Rabbit
Antigen Affinity Chromatography
RUO
26061, 56794, 85255
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8CHM7, Q9QXE0, Q9UJ83
Hacl1
HACL1 Fusion Protein Ag7830
150 μL
Primary
Human, Mouse, Rat