missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HACE1 Monoclonal Antibody (1854CT757.66.47)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537760
This item is not returnable.
View return policy
Description
HACE1 Monoclonal Antibody for Western Blot
E3 ubiquitin-protein ligase involved in Golgi membrane fusion and regulation of small GTPases. Acts as a regulator of Golgi membrane dynamics during the cell cycle: recruited to Golgi membrane by Rab proteins and regulates postmitotic Golgi membrane fusion. Acts by mediating ubiquitination during mitotic Golgi disassembly, ubiquitination serving as a signal for Golgi reassembly later, after cell division. Specifically interacts with GTP-bound RAC1, mediating ubiquitination and subsequent degradation of active RAC1, thereby playing a role in host defense against pathogens. May also act as a transcription regulator via its interaction with RARB.
Specifications
| HACE1 | |
| Monoclonal | |
| Unconjugated | |
| HACE1 | |
| 1700042J16Rik; A730034A22Rik; BC025474; E3 ubiquitin-protein ligase HACE1; hace 1; Hace1; HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1; HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1; HECT domain and ankyrin repeat-containing E3 ubiquitin-protein ligase 1; HECT-type E3 ubiquitin transferase HACE1; KIAA1320; SPPRS | |
| Mouse | |
| Protein G | |
| RUO | |
| 57531 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1854CT757.66.47 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8IYU2 | |
| HACE1 | |
| recombinant protein from the human region of human HACE1. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction