missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HACE1 Monoclonal Antibody (8D3), Biotin

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517702
This item is not returnable.
View return policy
Description
MA5-17702 targets Hace 1 in ELISA, IF, IHC (P), and WB applications and shows reactivity with Human samples. The MA5-17702 immunogen is mouse monoclonal antibodies raised against His-tagged recombinant full length Human HACE1.
E3 ubiquitin-protein ligase involved in Golgi membrane fusion and regulation of small GTPases. Acts as a regulator of Golgi membrane dynamics during the cell cycle: recruited to Golgi membrane by Rab proteins and regulates postmitotic Golgi membrane fusion. Acts by mediating ubiquitination during mitotic Golgi disassembly, ubiquitination serving as a signal for Golgi reassembly later, after cell division. Specifically interacts with GTP-bound RAC1, mediating ubiquitination and subsequent degradation of active RAC1, thereby playing a role in host defense against pathogens. May also act as a transcription regulator via its interaction with RARB.
Specifications
| HACE1 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
| Q8IYU2 | |
| HACE1 | |
| Mouse monoclonal antibodies raised against His-tagged recombinant full length Human HACE1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 8D3 | |
| Biotin | |
| HACE1 | |
| 1700042J16Rik; A730034A22Rik; BC025474; E3 ubiquitin-protein ligase HACE1; hace 1; Hace1; HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1; HECT domain and ankyrin repeat containing, E3 ubiquitin protein ligase 1; HECT domain and ankyrin repeat-containing E3 ubiquitin-protein ligase 1; HECT-type E3 ubiquitin transferase HACE1; KIAA1320; SPPRS | |
| Mouse | |
| Protein G | |
| RUO | |
| 57531 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction